Merck
  • Synthesis and biological evaluation of 7-substituted-1-(3-bromophenylamino)isoquinoline-4-carbonitriles as inhibitors of myosin light chain kinase and epidermal growth factor receptor.
Synthesis and biological evaluation of 7-substituted-1-(3-bromophenylamino)isoquinoline-4-carbonitriles as inhibitors of myosin light chain kinase and epidermal growth factor receptor.
Bioorganic & medicinal chemistry (2010-12-07)
Haridas B Rode, Martin L Sos, Christian Grütter, Stefanie Heynck, Jeffrey R Simard, Daniel Rauh
ABSTRACT
Here we present the synthesis and biological activity of a series of 7-substituted-1-(3-bromophenylamino)isoquinoline-4-carbonitriles as inhibitors of myosin light chain kinase (MLCK) and the epidermal growth factor receptor kinase (EGFR). The inhibitory effect of these molecules was found to be dependent on the nature of the substituents at the 7-position of the isoquinoline scaffold.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
K-252a, Ready Made Solution, from Nonomuraea longicatena, >98%