Merck
  • Macrophage migration inhibitory factor modulates cytokine expression in the trophoblast cell line HTR-8/SVneo.
Macrophage migration inhibitory factor modulates cytokine expression in the trophoblast cell line HTR-8/SVneo.
Reproduction, fertility, and development (2020-12-17)
Milica Jovanović Krivokuća, Ivana Stefanoska, Aleksandra Vilotić, Danica Ćujić, Svetlana Vrzić Petronijević, Ljiljana Vićovac
ABSTRACT
Extravillous trophoblasts are specific placental cells that invade the uterine stroma and spiral arteries modifying and adjusting them to pregnancy. Many pregnancy pathologies are associated with impairment of this process, including preeclampsia and intrauterine growth restriction, among others. Macrophage migration inhibitory factor (MIF) is a pleiotropic cytokine that is abundant at the fetomaternal interface. Previous results from our group showed that MIF participates in trophoblast invasion and modulates the expression of molecules known to mediate stromal and endovascular trophoblast invasion. In this study we investigated the possibility that MIF could act as a regulator of cytokines known to modulate trophoblast invasion using the normal extravillous trophoblast-derived cell line HTR-8/SVneo. Expression of trophoblast MIF was attenuated by MIF mRNA-specific small interfering RNAs. Cytokine expression was assessed at the mRNA and protein levels using real-time quantitative polymerase chain reaction and flow cytometry respectively. Knockdown of MIF led to a significant decrease in mRNA for IL-1β (IL1B) and IL-8 (CXCL8) and interleukin (IL)-8 protein. The addition of recombinant human MIF to cell culture medium increased IL-6 after 24h treatment and IL-6 and IL-8 after 72h treatment. Cell viability was not affected by MIF silencing or rhMIF treatment. The results of this study imply that at least some of the effects of MIF on trophoblast invasion could be mediated through autocrine or paracrine modulation of trophoblast cytokine production.
Sigma-Aldrich
MISSION® esiRNA, targeting human MIF
Pricing and availability is not currently available.