- Lignification in trees: indication of exclusive peroxidase participation.
Lignification in trees: indication of exclusive peroxidase participation.
Science (New York, N.Y.) (1973-04-20)
J M Harkin, J R Obst
PMID17816289
ABSTRACT
Syringaldazine did not turn purple on cross sections of tree branches or saplings or on cambial tissue cultures unless hydrogen peroxide was added; this indicated the absence of laccase but presence of peroxidase in lignifying cells. Peroxidase, therefore, apparently is the only en enzyme that polymerizes p-coumaryl alcohols to lignin in trees.