Merck
  • Messenger RNA expression and localization of xenin in the gastrointestinal tract in sheep.
Messenger RNA expression and localization of xenin in the gastrointestinal tract in sheep.
Domestic animal endocrinology (2020-08-17)
T Onaga, A Sakai, M Kajita, H Fukuda, Y Yasui, H Hayashi
ABSTRACT
The present study aimed to determine the primary sequence of ovine xenin and clarify the mRNA expression and peptide localization of xenin in the gastrointestinal tract in sheep. The colocalization of xenin and glucose-dependent insulinotropic polypeptide was also compared in the antrum and duodenum. Analysis of the nucleotide sequence of ovine xenin revealed a high degree (97.9%) of sequence homology of the sequence between sheep and cattle, and the amino acids sequence determined for ovine xenin coincided (100%) with that of other mammalian species. Real-time quantitative PCR for ovine xenin did not show regional difference in the mRNA expression ratio of xenin. In contrast to the real-time quantitative PCR results, anti-xenin positive cells were abundantly localized in the abomasal antrum (P < 0.01) and at a lesser amount in the duodenum, but no antixenin positive cells were observed in the other regions. Anti-xenin single-positive cells were in a majority in the abomasal antrum, whereas anti-xenin single-positive cells, and anti-GIP single-positive cells, and double-positive cells were even colocalized in the duodenum. These results suggest that abomasal antrum is a major source of xenin in the ovine gastrointestinal tract.
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Peptide YY porcine, ≥97% (HPLC), synthetic