Merck
  • Intermittent enzyme replacement therapy with recombinant human β-galactosidase prevents neuraminidase 1 deficiency
Intermittent enzyme replacement therapy with recombinant human β-galactosidase prevents neuraminidase 1 deficiency
The Journal of biological chemistry (2020-07-31)
Amanda R Luu, Cara Wong, Vishal Agrawal, Nathan Wise, Britta Handyside, Melanie J Lo, Glenn Pacheco, Jessica B Felix, Alexander Giaramita, Alessandra d'Azzo, Jon Vincelette, Sherry Bullens, Stuart Bunting, Terri M Christianson, Charles M Hague, Jonathan H LeBowitz, Gouri Yogalingam
ABSTRACT
Mutations in the galactosidase β 1 (GLB1) gene cause lysosomal β-galactosidase (β-Gal) deficiency and clinical onset of the neurodegenerative lysosomal storage disease, GM1 gangliosidosis. β-Gal and neuraminidase 1 (NEU1) form a multienzyme complex in lysosomes along with the molecular chaperone, protective protein cathepsin A (PPCA). NEU1 is deficient in the neurodegenerative lysosomal storage disease sialidosis, and its targeting to and stability in lysosomes strictly depend on PPCA. In contrast, β-Gal only partially depends on PPCA, prompting us to investigate the role that β-Gal plays in the multienzyme complex. Here, we demonstrate that β-Gal negatively regulates NEU1 levels in lysosomes by competitively displacing this labile sialidase from PPCA. Chronic cellular uptake of purified recombinant human β-Gal (rhβ-Gal) or chronic lentiviral-mediated GLB1 overexpression in GM1 gangliosidosis patient fibroblasts coincides with profound secondary NEU1 deficiency. A regimen of intermittent enzyme replacement therapy dosing with rhβ-Gal, followed by enzyme withdrawal, is sufficient to augment β-Gal activity levels in GM1 gangliosidosis patient fibroblasts without promoting NEU1 deficiency. In the absence of β-Gal, NEU1 levels are elevated in the GM1 gangliosidosis mouse brain, which are restored to normal levels following weekly intracerebroventricular dosing with rhβ-Gal. Collectively, our results highlight the need to carefully titrate the dose and dosing frequency of β-Gal augmentation therapy for GM1 gangliosidosis. They further suggest that intermittent intracerebroventricular enzyme replacement therapy dosing with rhβ-Gal is a tunable approach that can safely augment β-Gal levels while maintaining NEU1 at physiological levels in the GM1 gangliosidosis brain.
Sigma-Aldrich
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4-Methylumbelliferyl β-D-galactopyranoside, ≥99% (TLC)
Pricing and availability is not currently available.