Merck
  • Deep-learning-based three-dimensional label-free tracking and analysis of immunological synapses of CAR-T cells.
Deep-learning-based three-dimensional label-free tracking and analysis of immunological synapses of CAR-T cells.
eLife (2020-12-18)
Moosung Lee, Young-Ho Lee, Jinyeop Song, Geon Kim, YoungJu Jo, HyunSeok Min, Chan Hyuk Kim, YongKeun Park
ABSTRACT
The immunological synapse (IS) is a cell-cell junction between a T cell and a professional antigen-presenting cell. Since the IS formation is a critical step for the initiation of an antigen-specific immune response, various live-cell imaging techniques, most of which rely on fluorescence microscopy, have been used to study the dynamics of IS. However, the inherent limitations associated with the fluorescence-based imaging, such as photo-bleaching and photo-toxicity, prevent the long-term assessment of dynamic changes of IS with high frequency. Here, we propose and experimentally validate a label-free, volumetric, and automated assessment method for IS dynamics using a combinational approach of optical diffraction tomography and deep learning-based segmentation. The proposed method enables an automatic and quantitative spatiotemporal analysis of IS kinetics of morphological and biochemical parameters associated with IS dynamics, providing a new option for immunological research.
Sigma-Aldrich
Protamine sulfate, meets USP testing specifications
Pricing and availability is not currently available.