Merck
  • Protective effects of L-arginine on pulmonary oxidative stress and antioxidant defenses during exhaustive exercise in rats.
Protective effects of L-arginine on pulmonary oxidative stress and antioxidant defenses during exhaustive exercise in rats.
Acta pharmacologica Sinica (2005-07-26)
Wang-teng Lin, Suh-ching Yang, Kung-tung Chen, Chi-chang Huang, Ning-yuean Lee
ABSTRACT
To assess the effects of L-arginine (L-Arg) supplementation on pulmonary oxidative stress and antioxidant defenses in rats after exhaustive exercise. Rats were randomly divided into four groups: sedentary control (SC), sedentary control with L-Arg treatment (SC+Arg), exhaustive exercise with control diet (E) and exhaustive exercise with L-Arg treatment (E+Arg). Rats in groups SC+Arg and E+Arg received a 2% L-Arg diet. Rats in groups E and E+Arg underwent an exhaustive running test on a motorized treadmill. Pulmonary oxidative stress indices [xanthine oxidase (XO), myeloperoxidase (MPO), and malondialdehyde (MDA)] and antioxidant defense systems [superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPX), glutathione reductase (GR), and glutathione (GSH)] were investigated in this study. L-Arg supplementation significantly reduced exercise-induced elevations of XO and MPO activities in lung. L-Arg reversed the exercise-induced increase in SOD and GR activities, but increased CAT and GPX activities. L-Arg administration also significantly increased the GSH levels in plasma. L-Arg supplementation can prevent elevations of XO and MPO activities in the lung and favorably influence pulmonary antioxidant defense systems after exhaustive exercise.
Sigma-Aldrich
Glutathione Assay Kit
Pricing and availability is not currently available.