Merck
  • The temporal requirement for endothelin receptor-B signalling during neural crest development.
The temporal requirement for endothelin receptor-B signalling during neural crest development.
Nature (1999-12-11)
M K Shin, J M Levorse, R S Ingram, S M Tilghman
ABSTRACT
Endothelin receptor B (EDNRB) is a G-protein-coupled receptor with seven transmembrane domains which is required for the development of melanocytes and enteric neurons. Mice that are homozygous for a null mutation in the Ednrb gene are almost completely white and die as juveniles from megacolon. To determine when EDNRB signalling is required during embryogenesis, we have exploited the tetracycline-inducible system to generate strains of mice in which the endogenous Ednrb locus is under the control of the tetracycline-dependant transactivators tTa or rtTA. By using this system to express Ednrb at different stages of embryogenesis, we have determined that EDNRB is required during a restricted period of neural crest development between embryonic days 10 and 12.5. Moreover, our results imply that EDNRB is required for the migration of both melanoblasts and enteric neuroblasts.