Merck
  • Expression and Localization of BDNF/TrkB System in the Zebrafish Inner Ear.
Expression and Localization of BDNF/TrkB System in the Zebrafish Inner Ear.
International journal of molecular sciences (2020-08-19)
Antonino Germanà, Maria Cristina Guerrera, Rosaria Laurà, Maria Levanti, Marialuisa Aragona, Kamel Mhalhel, Germana Germanà, Giuseppe Montalbano, Francesco Abbate
ABSTRACT
Brain-derived neurotrophic factor (BDNF), a member of the neurotrophin family, is involved in multiple and fundamental functions of the central and peripheral nervous systems including sensory organs. Despite recent advances in knowledge on the functional significance of BDNF and TrkB in the regulation of the acoustic system of mammals, the localization of BDNF/TrkB system in the inner ear of zebrafish during development, is not well known. Therefore, the goal of the present study is to analyze the age-dependent changes using RT-PCR, Western Blot and single and double immunofluorescence of the BDNF and its specific receptor in the zebrafish inner ear. The results showed the mRNA expression and the cell localization of BDNF and TrkB in the hair cells of the crista ampullaris and in the neuroepithelium of the utricle, saccule and macula lagena, analyzed at different ages. Our results demonstrate that the BDNF/TrkB system is present in the sensory cells of the inner ear, during whole life. Therefore, this system might play a key role in the development and maintenance of the hair cells in adults, suggesting that the zebrafish inner ear represents an interesting model to study the involvement of the neurotrophins in the biology of sensory cells.
Sigma-Aldrich
Anti-Brain Derived Neurotrophic Factor Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.