Merck
  • The potent non-competitive mGlu1 receptor antagonist BAY 36-7620 differentially affects synaptic plasticity in area cornu ammonis 1 of rat hippocampal slices and impairs acquisition in the water maze task in mice.
The potent non-competitive mGlu1 receptor antagonist BAY 36-7620 differentially affects synaptic plasticity in area cornu ammonis 1 of rat hippocampal slices and impairs acquisition in the water maze task in mice.
Neuroscience (2008-10-04)
U H Schröder, T Müller, R Schreiber, A Stolle, W Zuschratter, D Balschun, R Jork, K G Reymann
ABSTRACT
In this study we evaluated the effects of the novel, potent non-competitive metabotropic glutamate receptor (mGluR) 1 antagonist (3aS,6aS)-6a-naphthalen-2-ylmethyl-5-methyliden-hexahydro-cyclopental[c]furan-1-on (BAY 36-7620) on different types of synaptic plasticity in the hippocampal cornu ammonis (CA) 1-region and on hippocampus-dependent spatial learning. After having confirmed the presence of mGluR1 in the hippocampal CA1 region of our rat strain by confocal microscopy, we tested the effects of BAY 36-7620 on: 1) long-term potentiation (LTP) induced by weak and strong stimulation; 2) 3,5-dihydroxyphenylglycine (DHPG, 30 microM)-induced depression of synaptic transmission; and 3) learning of the hidden platform version of the water maze by mice. BAY 36-7620 (10 microM) amplified LTP but, like the mGluR1 antagonists 7-hydroxyiminocyclopropan[b]chromen-1a-carboxylic acid ethyl ester (CPCCOEt, 10 microM) and 4-carboxyphenylglycine (4-CPG, 50 microM), diminished LTP at 1 microM. The mGluR5 antagonist 6-methyl-2-(phenylethynyl)-pyridine (MPEP, 10 microM) had no effect. BAY 36-7620 (10 microM) did not affect strong LTP. Thus, mGlu 1, but not mGlu 5, receptors modulate LTP elicited by weak stimulation in vitro. DHPG-induced depression of synaptic transmission was only marginally affected by BAY 36-7620 (1 microM) or 4-CPG (100 microM). In a mouse water maze study, BAY 36-7620 (10 mg/kg, i.v.) increased the escape latency and impaired water escape task acquisition during the first 4 days. Drug- and vehicle-treated groups showed comparable performance at day 5. Our data support a role for mGluR1 in LTP and in the acquisition of spatial memory.
Sigma-Aldrich
BAY 36-7620, ≥98% (HPLC)