Merck
  • Clinical Feasibility of Biofunctionalized Magnetic Nanoparticles for Detecting Multiple Cardiac Biomarkers in Emergency Chest Pain Patients.
Clinical Feasibility of Biofunctionalized Magnetic Nanoparticles for Detecting Multiple Cardiac Biomarkers in Emergency Chest Pain Patients.
Acta Cardiologica Sinica (2020-11-26)
Yen-Wen Wu, Yen-Ting Yeh, Chih-Cheng Wu, Chi-Lun Huang, Yi-Yao Chang, Chau-Chung Wu
ABSTRACT
The rapid diagnosis of acute myocardial infarction (AMI) is a clinical and operational priority in emergency departments. Serial serum levels of cardiac biomarkers play a crucial role in the evaluation of patients presenting with acute chest pain, so that an accurate and rapidly responsive assay of cardiac biomarkers is vital for emergency departments. Immunomagnetic reduction (IMR) has been developed for rapid and on-site assays with a small sample volume. IMR kits for three biomarkers [myoglobin, creatine kinase-MB (CK-MB), and troponin-I] have been developed by MagQu Co., Ltd., Taiwan (US patent: US20190072563A1). In this study, we examined correlations between IMR signals and biomarker concentrations. The measurement threshold of the IMR kits, dynamic ranges, interference tests in vitro, and reagent stability were tested. Clinical cases were included with serial IMR measurements to determine the time course and peak of IMR-measured cardiac biomarkers after AMI. The correlations between IMR signals and biomarker concentrations fitted well to logistic functions. The measurement thresholds of the IMR kits (1.03 × 10-8 ng/mL for myoglobin, 1.46 × 10-6 ng/mL for CK-MB, and 0.08 ng/mL for troponin-I) were much lower than the levels detected in the patients with AMI. There was no significant interference in vitro. The peak times of IMR-detected myoglobin, CK-MB, and troponin-I after AMI were 8.2 hours, 24.4 hours, and 24.7 hours, respectively. IMR is an accurate and sensitive on-site rapid assay for multiple cardiac biomarkers in vitro, and may play a role in the early diagnosis of AMI. Clinical trials are needed.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)