Merck
  • Beta-nerve growth factor stimulates spontaneous electrical activity of in vitro embryonic mouse GnRH neurons through a P75 mediated-mechanism.
Beta-nerve growth factor stimulates spontaneous electrical activity of in vitro embryonic mouse GnRH neurons through a P75 mediated-mechanism.
Scientific reports (2020-07-02)
Caroline Pinet-Charvet, Renaud Fleurot, Flavie Derouin-Tochon, Simon de Graaf, Xavier Druart, Guillaume Tsikis, Catherine Taragnat, Ana-Paula Teixeira-Gomes, Valérie Labas, Thierry Moreau, Xavier Cayla, Anne H Duittoz
ABSTRACT
The control of ovulation helps guarantee the success of reproduction and as such, contributes to the fitness of a species. In mammals, two types of ovulation are observed: induced and spontaneous ovulation. Recent work on camelids, that are induced ovulators, highlighted the role of a factor present in seminal plasma, beta Nerve Growth Factor (β-NGF), as the factor that triggers ovulation in a GnRH dependent manner. In the present work, we characterized alpaca β-NGF (aβ-NGF) and its 3D structure and compared it with human recombinant β-NGF (hβ-NGF). We showed that the β-NGF enriched fraction of alpaca semen and the human recombinant protein, both stimulated spontaneous electrical activity of primary GnRH neurons derived from mouse embryonic olfactory placodes. This effect was dose-dependent and mediated by p75 receptor signaling. P75 receptors were found expressed in vitro by olfactory ensheathing cells (OEC) in close association with GnRH neurons and in vivo by tanycytes in close vicinity to GnRH fibers in adult mouse. Altogether, these results suggested that β-NGF induced ovulation through an increase in GnRH secretion provoked by a glial dependent P75 mediated mechanism.
Sigma-Aldrich
Goat Anti-Mouse IgM Antibody, µ chain, Rhodamine conjugate, Chemicon®, from goat
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TrkA Antibody, Upstate®, from rabbit
Pricing and availability is not currently available.