Merck
  • Discovery of a Redox Thiol Switch: Implications for Cellular Energy Metabolism.
Discovery of a Redox Thiol Switch: Implications for Cellular Energy Metabolism.
Molecular & cellular proteomics : MCP (2020-03-07)
Xing-Huang Gao, Ling Li, Marc Parisien, Jing Wu, Ilya Bederman, Zhaofeng Gao, Dawid Krokowski, Steven M Chirieleison, Derek Abbott, Benlian Wang, Peter Arvan, Mark Cameron, Mark Chance, Belinda Willard, Maria Hatzoglou
ABSTRACT
The redox-based modifications of cysteine residues in proteins regulate their function in many biological processes. The gas molecule H2S has been shown to persulfidate redox sensitive cysteine residues resulting in an H2S-modified proteome known as the sulfhydrome. Tandem Mass Tags (TMT) multiplexing strategies for large-scale proteomic analyses have become increasingly prevalent in detecting cysteine modifications. Here we developed a TMT-based proteomics approach for selectively trapping and tagging cysteine persulfides in the cellular proteomes. We revealed the natural protein sulfhydrome of two human cell lines, and identified insulin as a novel substrate in pancreatic beta cells. Moreover, we showed that under oxidative stress conditions, increased H2S can target enzymes involved in energy metabolism by switching specific cysteine modifications to persulfides. Specifically, we discovered a Redox Thiol Switch, from protein S-glutathioinylation to S-persulfidation (RTSGS). We propose that the RTSGS from S-glutathioinylation to S-persulfidation is a potential mechanism to fine tune cellular energy metabolism in response to different levels of oxidative stress.
Sigma-Aldrich
Anti-CTH antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2