Merck
  • Thioredoxin negatively regulates p38 MAP kinase activation and IL-6 production by tumor necrosis factor-alpha.
Thioredoxin negatively regulates p38 MAP kinase activation and IL-6 production by tumor necrosis factor-alpha.
Biochemical and biophysical research communications (1999-05-18)
S Hashimoto, K Matsumoto, Y Gon, S Furuichi, S Maruoka, I Takeshita, K Hirota, J Yodoi, T Horie
ABSTRACT
We examined the regulatory role of a reduction/oxidation (redox) control protein, thioredoxin (TRX), in tumor necrosis factor-alpha (TNF-alpha)-induced p38 MAP kinase activation and p38 MAP kinase-mediated cytokine expression utilizing TRX-transfected murine L929 cells (TRX14). The results showed that TNF-alpha-induced p38 MAP kinase activation and interleukin-6 (IL-6) production by TRX 14 were less than those by the parental L cells and the control transfected L cells (Neo-1). SB 203580 as the specific inhibitor for p38 MAP kinase activity inhibited TNF-alpha-induced IL-6 production by the parental L cells, indicating that TNF-alpha-activated p38 MAP kinase regulates IL-6 production by the cell lines used in this study. These results showed that overexpression of TRX negatively regulates p38 MAP kinase activation and p38 MAP kinase-mediated IL-6 production by TNF-alpha-stimulated cells, indicating that TRX is critical for p38 MAP kinase activation which regulates cytokine expression.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)