Merck
  • 3-Hydroxyanthranilic acid inhibits PDK1 activation and suppresses experimental asthma by inducing T cell apoptosis.
3-Hydroxyanthranilic acid inhibits PDK1 activation and suppresses experimental asthma by inducing T cell apoptosis.
Proceedings of the National Academy of Sciences of the United States of America (2007-11-16)
Tomoko Hayashi, Ji-Hun Mo, Xing Gong, Cyprian Rossetto, Angela Jang, Lucinda Beck, Gregory I Elliott, Irina Kufareva, Ruben Abagyan, David H Broide, Jongdae Lee, Eyal Raz
ABSTRACT
3-Hydroxyanthranilic acid (HAA), a compound generated during tryptophan metabolism initiated by indoleamine 2,3-dioxygenase, is known to induce T cell death, but its molecular target is not known. Here we report that HAA inhibits NF-kappaB activation upon T cell antigen receptor engagement by specifically targeting PDK1. Inhibition of NF-kappaB by HAA leads to dysfunction and cell death of activated Th2 cells, which in turn suppresses experimental asthma. Inhibition of NF-kappaB and induction of apoptosis is specific to CD4 T cells because HAA does not inhibit NF-kappaB activation or induce cell death upon Toll-like receptor 4 stimulation in dendritic cells. Thus, HAA is a natural inhibitor that restrains T cell expansion and activation.
Sigma-Aldrich
2,3-Pyridinedicarboxylic acid, 99%