Merck
  • Cytosolic phospholipase A(2) regulates golgi structure and modulates intracellular trafficking of membrane proteins.
Cytosolic phospholipase A(2) regulates golgi structure and modulates intracellular trafficking of membrane proteins.
The Journal of clinical investigation (2000-10-18)
G J Choukroun, V Marshansky, C E Gustafson, M McKee, R J Hajjar, A Rosenzweig, D Brown, J V Bonventre
ABSTRACT
The Golgi complex and the trans-Golgi network are critical cellular organelles involved in the endocytic and biosynthetic pathways of protein trafficking. Lipids have been implicated in the regulation of membrane-protein trafficking, vesicular fusion, and targeting. We have explored the role of cytosolic group IV phospholipase A(2) (cPLA(2)) in membrane-protein trafficking in kidney epithelial cells. Adenoviral expression of cPLA(2) in LLC-PK(1) kidney epithelial cells prevents constitutive trafficking to the plasma membrane of an aquaporin 2-green fluorescent protein chimera, with retention of the protein in the rough endoplasmic reticulum. Plasma membrane Na(+)-K(+)-ATPase alpha-subunit localization is markedly reduced in cells expressing cPLA(2), whereas the trafficking of a Cl(-)/HCO(3)(-) anion exchanger to the plasma membrane is not altered in these cells. Expression of cPLA(2) results in dispersion of giantin and beta-COP from their normal, condensed Golgi localization, and in marked disruption of the Golgi cisternae. cPLA(2) is present in Golgi fractions from noninfected LLC-PK(1) cells and rat kidney cortex. The distribution of tubulin and actin was not altered by cPLA(2), indicating that the microtubule and actin cytoskeleton remain intact. Total cellular protein synthesis is unaffected by the increase in cPLA(2) activity. Thus cPLA(2) plays an important role in determining Golgi architecture and selective control of constitutive membrane-protein trafficking in renal epithelial cells.
Sigma-Aldrich
Monoclonal Anti-β-COP antibody produced in mouse, clone M3A5, ascites fluid
Pricing and availability is not currently available.