Merck
  • Distinct Polysaccharide Utilization Determines Interspecies Competition between Intestinal Prevotella spp.
Distinct Polysaccharide Utilization Determines Interspecies Competition between Intestinal Prevotella spp.
Cell host & microbe (2020-10-29)
Eric J C Gálvez, Aida Iljazovic, Lena Amend, Till Robin Lesker, Thibaud Renault, Sophie Thiemann, Lianxu Hao, Urmi Roy, Achim Gronow, Emmanuelle Charpentier, Till Strowig
ABSTRACT
Prevotella spp. are a dominant bacterial genus within the human gut. Multiple Prevotella spp. co-exist in some individuals, particularly those consuming plant-based diets. Additionally, Prevotella spp. exhibit variability in the utilization of diverse complex carbohydrates. To investigate the relationship between Prevotella competition and diet, we isolated Prevotella species from the mouse gut, analyzed their genomes and transcriptomes in vivo, and performed competition experiments between species in mice. Diverse dominant Prevotella species compete for similar metabolic niches in vivo, which is linked to the upregulation of specific polysaccharide utilization loci (PULs). Complex plant-derived polysaccharides are required for Prevotella spp. expansion, with arabinoxylans having a prominent impact on species abundance. The most dominant Prevotella species encodes a specific tandem-repeat trsusC/D PUL that enables arabinoxylan utilization and is conserved in human Prevotella copri strains, particularly among those consuming a vegan diet. These findings suggest that efficient (arabino)xylan-utilization is a factor contributing to Prevotella dominance.
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Supelco
Pyridoxine Hydrochloride (B6), analytical standard
Sigma-Aldrich
Thiamine hydrochloride, reagent grade, ≥99% (HPLC)