Merck
  • Towards the scale up of tissue engineered intervertebral discs for clinical application.
Towards the scale up of tissue engineered intervertebral discs for clinical application.
Acta biomaterialia (2018-02-11)
Sarah E Gullbrand, Dong Hwa Kim, Edward Bonnevie, Beth G Ashinsky, Lachlan J Smith, Dawn M Elliott, Robert L Mauck, Harvey E Smith
ABSTRACT
Replacement of the intervertebral disc with a viable, tissue-engineered construct that mimics native tissue structure and function is an attractive alternative to fusion or mechanical arthroplasty for the treatment of disc pathology. While a number of engineered discs have been developed, the average size of these constructs remains a fraction of the size of human intervertebral discs. In this study, we fabricated medium (3 mm height × 10 mm diameter) and large (6 mm height × 20 mm diameter) sized disc-like angle ply structures (DAPS), encompassing size scales from the rabbit lumbar spine to the human cervical spine. Maturation of these engineered discs was evaluated over 15 weeks in culture by quantifying cell viability and metabolic activity, construct biochemical content, MRI T2 values, and mechanical properties. To assess the performance of the DAPS in the in vivo space, pre-cultured DAPS were implanted subcutaneously in athymic rats for 5 weeks. Our findings show that both sized DAPS matured functionally and compositionally during in vitro culture, as evidenced by increases in mechanical properties and biochemical content over time, yet large DAPS under-performed compared to medium DAPS. Subcutaneous implantation resulted in reductions in NP cell viability and GAG content at both size scales, with little effect on AF biochemistry or metabolic activity. These findings demonstrate that engineered discs at large size scales will mature during in vitro culture, however, future work will need to address the challenges of reduced cell viability and heterogeneous matrix distribution throughout the construct. This work establishes, for the first time, tissue-engineered intervertebral discs for total disc replacement at large, clinically relevant length scales. Clinical translation of tissue-engineered discs will offer an alternative to mechanical disc arthroplasty and fusion procedures, and may contribute to a paradigm shift in the clinical care for patients with disc pathology and associated axial spine and neurogenic extremity pain.