Merck
  • Rhizobacterial biofilm and plant growth promoting trait enhancement by organic acids and sugars.
Rhizobacterial biofilm and plant growth promoting trait enhancement by organic acids and sugars.
Biofouling (2020-11-06)
Jishma Panichikkal, Radhakrishnan Edayileveetil Krishnankutty
ABSTRACT
In the study,Pseudomonas sp. K6 and Pseudomonas monteilii were found to form an enhanced biofilm when cultured in the presence of organic acids and sugars. Here, the highest biofilm could be observed for Pseudomonas sp. K6 (3.08 ± 0.13) and P. monteilii (1.99 ± 0.12) when cultured in presence of 10 µM malic acid. However, maximum production of indole 3 acetic acid (IAA) was observed with 25 µM succinic acid treatment for Pseudomonas sp. K6 (24.33 ± 0.57 µg ml-1) and with 25 µM galactose for P. monteilii (20 ± 0.0 µg ml-1). At the same time, Pseudomonas sp. K6 solubilized the highest quantity of phosphate in the presence of 50 µM citric acid (21.33 ± 0.0 µM) and P. monteilii was observed to produce 32.66 ± 1.25 µM soluble phosphate in the presence of 10 µM galactose. The results of the study demonstrate the role of organic acids and sugars in the enhancement of biofilm formation, IAA production and phosphate solubilization in selected Pseudomonas spp. and highlight the potential use of rhizobacteria in conjugation with supplement for the agricultural applications.
Sigma-Aldrich
Malachite Green Phosphate Assay Kit, sufficient for 2500 colorimetric tests
Supelco
Malachite Green chloride, analytical standard