Merck
  • Rice NIN-LIKE PROTEIN 1 rapidly responds to nitrogen deficiency and improves yield and nitrogen use efficiency.
Rice NIN-LIKE PROTEIN 1 rapidly responds to nitrogen deficiency and improves yield and nitrogen use efficiency.
Journal of experimental botany (2020-06-26)
Alamin Alfatih, Jie Wu, Zi-Sheng Zhang, Jin-Qiu Xia, Sami Ullah Jan, Lin-Hui Yu, Cheng-Bin Xiang
ABSTRACT
Nitrogen (N) is indispensable for crop growth and yield, but excessive agricultural application of nitrogenous fertilizers has generated severe environmental problems. A desirable and economical solution to cope with these issues is to improve crop nitrogen use efficiency (NUE). Plant NUE has been a focal point of intensive research worldwide, yet much still has to be learned about its genetic determinants and regulation. Here, we show that rice (Oryza sativa L.) NIN-LIKE PROTEIN 1 (OsNLP1) plays a fundamental role in N utilization. OsNLP1 protein localizes in the nucleus and its transcript level is rapidly induced by N starvation. Overexpression of OsNLP1 improves plant growth, grain yield, and NUE under different N conditions, while knockout of OsNLP1 impairs grain yield and NUE under N-limiting conditions. OsNLP1 regulates nitrate and ammonium utilization by cooperatively orchestrating multiple N uptake and assimilation genes. Chromatin immunoprecipitation and yeast one-hybrid assays showed that OsNLP1 can directly bind to the promoter of these genes to activate their expression. Therefore, our results demonstrate that OsNLP1 is a key regulator of N utilization and represents a potential target for improving NUE and yield in rice.
Sigma-Aldrich
Potassium nitrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Ammonium-15N chloride, ≥98 atom % 15N, ≥99% (CP)
Sigma-Aldrich
Potassium nitrate-15N, 98 atom % 15N