Merck
  • Performance of four chambers to measure sperm concentration: results from an external quality assurance programme.
Performance of four chambers to measure sperm concentration: results from an external quality assurance programme.
Reproductive biomedicine online (2020-08-28)
Emily Zuvela, Phillip Matson
ABSTRACT
What are the changes in the use of four types of counting chambers by laboratories enrolled in an Australian-based external quality assurance programme, and what are their accuracy and precision? Samples of latex beads of known concentration up to 20 × 106/ml were distributed quarterly to enrolled laboratories over a 12-year period. The results of each distribution were then used to calculate a bias relative to the target value as an indicator of accuracy and a coefficient of variation to indicate the level of precision. The proportion of laboratories in 2007-2008 using improved Neubauer haemocytometers (44%), Makler® (9%) and Vetriplast chambers (19%) remained constant in 2018-2019, unlike Kova chamber users (20%), which reduced. The mean (range) bias of improved Neubauer haemocytometers (-2.8% [-22.5 to +32.0%]) was less than Makler® chambers (+17.0% [-2.9 to +41.2%]), Kova chambers (+33.9% [0.0 to 115.0%]) and Vetriplast chambers (+47.9% [0.0 to 170.0%]). The coefficient of variation of improved Neubauer haemocytometers (14.6% [8.7 to 25.0%]) was less than both Vetriplast (20.7% [8.8 to 36.4%]) and Makler® (24.1% [13.6 to 48.6%]) and Kova chambers (35.5% [15.9 to 123.0%]). The improved Neubauer haemocytometer has been shown to be superior in accuracy and precision to the Makler®, Kova and Vetriplast chambers in their estimation of concentrations up to 20 × 106/ml. Users of Makler® chambers, specifically designed for counting spermatozoa, should take care to monitor the performance of their own chambers, whereas Kova and Vetriplast chambers (designed for microscopic urinalysis) should not be used.
Product Description
Latex beads, styrene divinylbenzene, aqueous suspension, 6.4 μm mean particle size