Merck
  • Design, Synthesis, and Characterization of 4-Undecylpiperidine-2-carboxamides as Positive Allosteric Modulators of the Serotonin (5-HT) 5-HT2C Receptor.
Design, Synthesis, and Characterization of 4-Undecylpiperidine-2-carboxamides as Positive Allosteric Modulators of the Serotonin (5-HT) 5-HT2C Receptor.
Journal of medicinal chemistry (2018-04-06)
Christopher T Wild, Joanna M Miszkiel, Eric A Wold, Claudia A Soto, Chunyong Ding, Rachel M Hartley, Mark A White, Noelle C Anastasio, Kathryn A Cunningham, Jia Zhou
ABSTRACT
An impaired signaling capacity of the serotonin (5-HT) 5-HT2C receptor (5-HT2CR) has been implicated in the neurobehavioral processes that promote relapse vulnerability in cocaine use disorder (CUD). Restoration of the diminished 5-HT2CR signaling through positive allosteric modulation presents a novel therapeutic approach. Several new molecules with the 4-alkylpiperidine-2-carboxamide scaffold were designed, synthesized, and pharmacologically evaluated, leading to the discovery of selective 5-HT2CR positive allosteric modulators (PAMs). Compound 16 (CYD-1-79) potentiated 5-HT-evoked intracellular calcium release in cells stably expressing the human 5-HT2CR but not the 5-HT2AR cells. A topographically distinct allosteric site was identified based on the newly solved 5-HT2CR structure. Compound 16 modulated 5-HT2CR-mediated spontaneous ambulation, partially substituted for the training dose of the 5-HT2CR agonist WAY163909, synergized with a low dose of WAY163909 to substitute fully for the stimulus effects of WAY163909, and attenuated relapse vulnerability as assessed in a rodent self-administration model, indicating its therapeutic promise for CUD.
Sigma-Aldrich
WAY-163909 hydrochloride, ≥98% (HPLC)