Merck
  • Neuroprotective efficacy of poly-arginine R18 and NA-1 (TAT-NR2B9c) peptides following transient middle cerebral artery occlusion in the rat.
Neuroprotective efficacy of poly-arginine R18 and NA-1 (TAT-NR2B9c) peptides following transient middle cerebral artery occlusion in the rat.
Neuroscience research (2016-09-19)
Diego Milani, Jane L Cross, Ryan S Anderton, David J Blacker, Neville W Knuckey, Bruno P Meloni
ABSTRACT
We examined the efficacy of R18 in a transient MCAO model and compared its effectiveness to the well-characterized neuroprotective NA-1 peptide. R18 and NA-1 peptides were administered intravenously (30, 100, 300, 1000nmol/kg), 60min after the onset of 90min of MCAO. Infarct volume, cerebral swelling and functional outcomes (neurological score, adhesive tape and rota-rod) were measured 24h after MCAO. R18 reduced total infarct volume by 35.1% (p=0.008), 24.8% (p=0.059), 12.2% and 9.6% for the respective 1000 to 30nmol/kg doses, while the corresponding doses of NA-1 reduced lesion volume by 26.1% (p=0.047), 16.6%, 16.5% and 7%, respectively. R18 also reduced hemisphere swelling by between 46.1% (1000 and 300nmol/kg; p=0.009) and 24.4% (100nmol/kg; p=0.066), while NA-1 reduced swelling by 25.7% (1000nmol/kg; p=0.054). In addition, several R18 and NA-1 treatment groups displayed a significant improvement in at least one parameter of the adhesive tape test. These results confirm the neuroprotective properties of R18, and suggest that the peptide is a more effective neuroprotective agent than NA-1. This provides strong justification for the continuing development of R18 as a neuroprotective treatment for stroke.