Merck
  • Pathogenesis of Intradermal Staphylococcal Infections: Rabbit Experimental Approach to Natural Staphylococcus aureus Skin Infections.
Pathogenesis of Intradermal Staphylococcal Infections: Rabbit Experimental Approach to Natural Staphylococcus aureus Skin Infections.
The American journal of pathology (2020-03-24)
Asunción Muñoz-Silvestre, Mariola Penadés, Laura Selva, Sara Pérez-Fuentes, Elena Moreno-Grua, Ana García-Quirós, Juan J Pascual, Alberto Arnau-Bonachera, Agustín Barragán, Juan M Corpa, David Viana
ABSTRACT
Despite the enormous efforts made to achieve effective tools that fight against Staphylococcus aureus, the results have not been successful. This failure may be due to the absence of truly representative experimental models. To overcome this deficiency, the present work describes and immunologically characterizes the infection for 28 days, in an experimental low-dose (300 colony-forming units) intradermal model of infection in rabbits, which reproduces the characteristic staphylococcal abscess. Surprisingly, when mutant strains in the genes involved in virulence (JΔagr, JΔcoaΔvwb, JΔhla, and JΔpsmα) were inoculated, no strong effect on the severity of lesions was observed, unlike other models that use high doses of bacteria. The inoculation of a human rabbitized (FdltBr) strain demonstrated its capacity to generate a similar inflammatory response to a wild-type rabbit strain and, therefore, validated this model for conducting these experimental studies with human strains. To conclude, this model proved reproducible and may be an option of choice to check both wild-type and mutant strains of different origins.
Sigma-Aldrich
Mouse Anti-Rabbit light chain, HRP conjugate Antibody, Chemicon®, from mouse
Pricing and availability is not currently available.