Merck
  • Developmental Relationships of Four Exhausted CD8+ T Cell Subsets Reveals Underlying Transcriptional and Epigenetic Landscape Control Mechanisms.
Developmental Relationships of Four Exhausted CD8+ T Cell Subsets Reveals Underlying Transcriptional and Epigenetic Landscape Control Mechanisms.
Immunity (2020-05-13)
Jean-Christophe Beltra, Sasikanth Manne, Mohamed S Abdel-Hakeem, Makoto Kurachi, Josephine R Giles, Zeyu Chen, Valentina Casella, Shin Foong Ngiow, Omar Khan, Yinghui Jane Huang, Patrick Yan, Kito Nzingha, Wei Xu, Ravi K Amaravadi, Xiaowei Xu, Giorgos C Karakousis, Tara C Mitchell, Lynn M Schuchter, Alexander C Huang, E John Wherry
ABSTRACT
CD8+ T cell exhaustion is a major barrier to current anti-cancer immunotherapies. Despite this, the developmental biology of exhausted CD8+ T cells (Tex) remains poorly defined, restraining improvement of strategies aimed at "re-invigorating" Tex cells. Here, we defined a four-cell-stage developmental framework for Tex cells. Two TCF1+ progenitor subsets were identified, one tissue restricted and quiescent and one more blood accessible, that gradually lost TCF1 as it divided and converted to a third intermediate Tex subset. This intermediate subset re-engaged some effector biology and increased upon PD-L1 blockade but ultimately converted into a fourth, terminally exhausted subset. By using transcriptional and epigenetic analyses, we identified the control mechanisms underlying subset transitions and defined a key interplay between TCF1, T-bet, and Tox in the process. These data reveal a four-stage developmental hierarchy for Tex cells and define the molecular, transcriptional, and epigenetic mechanisms that could provide opportunities to improve cancer immunotherapy.
Sigma-Aldrich
Percoll®, Cytiva 17-0891-01, pack of 1 L
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein