Merck
  • PTPN2 phosphatase deletion in T cells promotes anti-tumour immunity and CAR T-cell efficacy in solid tumours.
PTPN2 phosphatase deletion in T cells promotes anti-tumour immunity and CAR T-cell efficacy in solid tumours.
The EMBO journal (2019-12-06)
Florian Wiede, Kun-Hui Lu, Xin Du, Shuwei Liang, Katharina Hochheiser, Garron T Dodd, Pei K Goh, Conor Kearney, Deborah Meyran, Paul A Beavis, Melissa A Henderson, Simone L Park, Jason Waithman, Sheng Zhang, Zhong-Yin Zhang, Jane Oliaro, Thomas Gebhardt, Phillip K Darcy, Tony Tiganis
ABSTRACT
Although adoptive T-cell therapy has shown remarkable clinical efficacy in haematological malignancies, its success in combating solid tumours has been limited. Here, we report that PTPN2 deletion in T cells enhances cancer immunosurveillance and the efficacy of adoptively transferred tumour-specific T cells. T-cell-specific PTPN2 deficiency prevented tumours forming in aged mice heterozygous for the tumour suppressor p53. Adoptive transfer of PTPN2-deficient CD8+ T cells markedly repressed tumour formation in mice bearing mammary tumours. Moreover, PTPN2 deletion in T cells expressing a chimeric antigen receptor (CAR) specific for the oncoprotein HER-2 increased the activation of the Src family kinase LCK and cytokine-induced STAT-5 signalling, thereby enhancing both CAR T-cell activation and homing to CXCL9/10-expressing tumours to eradicate HER-2+ mammary tumours in vivo. Our findings define PTPN2 as a target for bolstering T-cell-mediated anti-tumour immunity and CAR T-cell therapy against solid tumours.
Sigma-Aldrich
Mouse IgG2a ELISA Kit
Sigma-Aldrich
MISSION® esiRNA, targeting human PTPN2