Merck
  • Receptor-driven, multimodal mapping of cortical areas in the macaque monkey intraparietal sulcus.
Receptor-driven, multimodal mapping of cortical areas in the macaque monkey intraparietal sulcus.
eLife (2020-07-03)
Meiqi Niu, Daniele Impieri, Lucija Rapan, Thomas Funck, Nicola Palomero-Gallagher, Karl Zilles
ABSTRACT
The intraparietal sulcus (IPS) is structurally and functionally heterogeneous. We performed a quantitative cyto-/myelo- and receptor architectonical analysis to provide a multimodal map of the macaque IPS. We identified 17 cortical areas, including novel areas PEipe, PEipi (external and internal subdivisions of PEip), and MIPd. Multivariate analyses of receptor densities resulted in a grouping of areas based on the degree of (dis)similarity of their receptor architecture: a cluster encompassing areas located in the posterior portion of the IPS and associated mainly with the processing of visual information, a cluster including areas found in the anterior portion of the IPS and involved in sensorimotor processing, and an 'intermediate' cluster of multimodal association areas. Thus, differences in cyto-/myelo- and receptor architecture segregate the cortical ribbon within the IPS, and receptor fingerprints provide novel insights into the relationship between the structural and functional segregation of this brain region in the macaque monkey.
Sigma-Aldrich
Zinc isopropylsulfinate, 95%