Merck
  • Artesunate exerts anti-prolactinoma activity by inhibiting mitochondrial metabolism and inducing apoptosis.
Artesunate exerts anti-prolactinoma activity by inhibiting mitochondrial metabolism and inducing apoptosis.
Annals of translational medicine (2020-08-15)
Weiyu Zhang, Qiu Du, Piaopiao Bian, Zheng Xiao, Xin Wang, Yajuan Feng, Hou Feng, Ziyan Zhu, Nailin Gao, Diming Zhu, Xiang Fan, Yonghong Zhu
ABSTRACT
Prolactinoma is the most common hormone-secreting pituitary adenoma. Dopamine receptor agonists (DAs) are effective in reducing prolactin levels and tumor mass, but some prolactinoma patients are resistant to DAs. Treating patients with DA-resistant prolactinoma is challenging. In this study, we examined the anti-prolactinoma effect of artesunate (ART), a potential new treatment option for prolactinoma, and its mechanism of action. Cell Counting Kit-8 (CCK8) and flow cytometry were used to detect the effect of ART on the proliferation, cycle, and apoptosis of rat pituitary adenoma cell line MMQ. The subcellular localization of ART was observed using confocal fluorescence microscopy. The JC-1 mitochondrial membrane potential (MMP) detection and Seahorse assays were used to detect the effect of ART on mitochondrial function. Real-time quantitative polymerase chain reaction (RT-qPCR) and Western blot analysis were used to detect the effect of ART on the expression of prolactin (PRL) and apoptosis-related proteins. A mouse xenograft model of prolactinoma was used to detect the inhibitory effect of ART on MMQ in vivo. ART specifically inhibited MMQ proliferation and PRL synthesis, induced G0/G1 phase arrest and apoptosis in vitro. ART accumulated in the mitochondria of MMQ cells, inhibiting mitochondrial respiratory function and mediating apoptosis through the mitochondrial pathway. ART also inhibited proliferation and activated the apoptosis of MMQ cells in vivo. ART has a strong inhibitory effect on prolactinoma both in vitro and in vivo, and its effects rely on high MMP to inhibit mitochondrial metabolism and induce apoptosis. Our results provide evidence for ART as a candidate drug for the treatment of prolactinoma.
Sigma-Aldrich
Artesunate, from Artemisia annua
Pricing and availability is not currently available.
Sigma-Aldrich
JC-1, powder or solid (Crystals)
Pricing and availability is not currently available.