Merck
  • MicroRNA-621 inhibits the growth of gastric cancer cells by targeting SYF2.
MicroRNA-621 inhibits the growth of gastric cancer cells by targeting SYF2.
Archives of biochemistry and biophysics (2020-05-18)
Youmao Tao, Yue Zhao, Yujiao Peng, Xinrui Ma, Caixia Sun, Kaicheng Xu
ABSTRACT
Increasing evidence indicates that aberrantly expressed microRNAs play a role in tumorigenesis and progression of gastric cancer. Recently, a novel cancer-related microRNA, miR-621, was found to be involved in cancer pathogenesis. However, the precise molecular mechanisms underlying the oncogenic activity of miR-621 remain unclear and require further investigation. In the current study, we demonstrate that miR-621 expression is downregulated in gastric cancer tissues and cell lines, and its reduction is associated with malignant clinical features including tumor size, lymph node metastasis, tumor-node-metastasis stage and poor prognosis. Functional studies involving gain- and loss-of-function experiments revealed that miR-621 represses cell viability, colony formation, cell cycle progression and proliferation in vitro, and miR-621 overexpression inhibited tumor growth in a gastric cancer xenograft model. SYF2 was identified as a direct target gene of miR-621 in gastric cancer. MiR-621 directly interacts with the SYF2 3'-UTR and post-transcriptionally repressed SYF2 expression in gastric cancer cells. SYF2 was significantly overexpressed in gastric cancer tissues and negatively correlated with miR-621 expression. Moreover, inhibition of SYF2 expression reversed the effects of miR-621 loss in gastric cancer cells. SYF2 overexpression was similar to that induced by miR-621 loss in gastric cancer. Taken together, these studies suggest that miR-621 may be a viable therapeutic target in gastric cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human SYF2
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder