Merck
  • Genotoxicity of aldehyde mixtures: profile of exocyclic DNA-adducts as a biomarker of exposure to tobacco smoke.
Genotoxicity of aldehyde mixtures: profile of exocyclic DNA-adducts as a biomarker of exposure to tobacco smoke.
Toxicology letters (2020-05-23)
Héléna Alamil, Laurence Galanti, Natacha Heutte, Marie Van Der Schueren, Zeina Dagher, Mathilde Lechevrel
ABSTRACT
Electrophilic compounds present in humans, originating from endogenous processes or pollutant exposures, pose a risk to health though their reaction with nucleophilic sites in protein and DNA. Among this chemical class, aldehydes are mainly present in indoor air and they can also be produced by endogenous lipid peroxidation arising from oxidative stress. Known to be very reactive, aldehydes have the ability to form exocyclic adducts to DNA that, for the most if not repaired correctly, are mutagenic and by consequence potential agents involved in carcinogenesis. The aim of this work was to establish profiles of exocyclic DNA adducts induced by aldehyde mixtures, which could ultimately be considered as a genotoxic marker of endogenous and environmental aldehyde exposure. Adducts were quantified by an accurate, sensitive and validated ultra high performance liquid chromatography-electrospray ionization analytical method coupled to mass spectrometry in the tandem mode (UHPLC-ESI-MS/MS). We simultaneously measured nine exocyclic DNA adducts generated during the exposure in vitro of calf thymus DNA to different concentrations of each aldehyde along, as well as, to an equimolar mixture of these aldehydes. This approach has enabled us to establish dose-response relationships that allowed displaying the specific reactivity of aldehydes towards corresponding adducts formation. Profiles of these adducts determined in DNA of current smokers and non-smokers blood samples supported these findings. These first results are encouraging to explore genotoxicity induced by aldehyde mixtures and can furthermore be used as future reference for adductomic approaches.
Sigma-Aldrich
HNE-DMA, hexane solution, ≥85% (GC)