Merck
  • D1-D2 dopamine receptor heterooligomers with unique pharmacology are coupled to rapid activation of Gq/11 in the striatum.
D1-D2 dopamine receptor heterooligomers with unique pharmacology are coupled to rapid activation of Gq/11 in the striatum.
Proceedings of the National Academy of Sciences of the United States of America (2006-12-30)
Asim J Rashid, Christopher H So, Michael M C Kong, Teresa Furtak, Mufida El-Ghundi, Regina Cheng, Brian F O'Dowd, Susan R George
ABSTRACT
We demonstrate a heteromeric D1-D2 dopamine receptor signaling complex in brain that is coupled to Gq/11 and requires agonist binding to both receptors for G protein activation and intracellular calcium release. The D1 agonist SKF83959 was identified as a specific agonist for the heteromer that activated Gq/11 by functioning as a full agonist for the D1 receptor and a high-affinity partial agonist for a pertussis toxin-resistant D2 receptor within the complex. We provide evidence that the D1-D2 signaling complex can be more readily detected in mice that are 8 months in age compared with animals that are 3 months old, suggesting that calcium signaling through the D1-D2 dopamine receptor complex is relevant for function in the postadolescent brain. Activation of Gq/11 through the heteromer increases levels of calcium/calmodulin-dependent protein kinase IIalpha in the nucleus accumbens, unlike activation of Gs/olf-coupled D1 receptors, indicating a mechanism by which D1-D2 dopamine receptor complexes may contribute to synaptic plasticity.
Sigma-Aldrich
R(+)-SKF-81297 hydrobromide, ≥98% (HPLC), solid
Pricing and availability is not currently available.