Merck
  • Overexpression, crystallization and preliminary X-ray crystallographic analysis of the C-terminal cytosolic domain of mouse anoctamin 1.
Overexpression, crystallization and preliminary X-ray crystallographic analysis of the C-terminal cytosolic domain of mouse anoctamin 1.
Acta crystallographica. Section F, Structural biology and crystallization communications (2011-11-22)
Sang Ho Park, Ho Kyung Chung, Do Jin Kim, Mi Ra Han, Mi Seul Park, Uhtaek Oh, Hyun Jung Kim, Byung Woo Han
ABSTRACT
Transmembrane protein 16A (TMEM16A, also known as anoctamin 1; ANO1) is a bona fide Ca(2+)-activated chloride channel that is activated by intracellular Ca(2+)- and Ca(2+)-mobilizing stimuli and plays important roles in a variety of physiological functions. To elucidate the structural features of ANO1, structural analysis of the C-terminal cytosolic domain of mouse ANO1 (mANO1-CTD) was initiated. mANO1-CTD was overexpressed in Escherichia coli and was crystallized at 297 K using a reservoir solution consisting of 0.2 M sodium acetate trihydrate, 0.1 M Tris-HCl pH 8.5 and 30%(w/v) PEG 4000. X-ray diffraction data were collected to 2.3 Å resolution. The crystals belonged to the orthorhombic space group P2(1)2(1)2(1), with unit-cell parameters a = 73.96, b = 103.73, c = 114.71 Å. If it is assumed that eight copies of a monomer molecule are present in the crystallographic asymmetric unit, the crystal volume per protein mass (V(M)) is 2.38 Å(3) Da(-1) and the solvent content is 48.38%. Attempts to solve the structure of mANO1-CTD by the MAD method using selenomethionine-labelled mANO1-CTD or heavy-atom-derivatized crystals are in progress.
Sigma-Aldrich
Sodium chloride physiological solution, BioUltra, tablet
Pricing and availability is not currently available.