Merck
  • Efficiency enhancement in low-bandgap polymer solar cells by processing with alkane dithiols.
Efficiency enhancement in low-bandgap polymer solar cells by processing with alkane dithiols.
Nature materials (2007-05-29)
J Peet, J Y Kim, N E Coates, W L Ma, D Moses, A J Heeger, G C Bazan
ABSTRACT
High charge-separation efficiency combined with the reduced fabrication costs associated with solution processing and the potential for implementation on flexible substrates make 'plastic' solar cells a compelling option for tomorrow's photovoltaics. Attempts to control the donor/acceptor morphology in bulk heterojunction materials as required for achieving high power-conversion efficiency have, however, met with limited success. By incorporating a few volume per cent of alkanedithiols in the solution used to spin-cast films comprising a low-bandgap polymer and a fullerene derivative, the power-conversion efficiency of photovoltaic cells (air-mass 1.5 global conditions) is increased from 2.8% to 5.5% through altering the bulk heterojunction morphology. This discovery can potentially enable morphological control in bulk heterojunction materials where thermal annealing is either undesirable or ineffective.
Sigma-Aldrich
PCPDTBT, average Mw 7,000-20,000
Pricing and availability is not currently available.