Merck
  • Lidocaine and ropivacaine, but not bupivacaine, demethylate deoxyribonucleic acid in breast cancer cells in vitro.
Lidocaine and ropivacaine, but not bupivacaine, demethylate deoxyribonucleic acid in breast cancer cells in vitro.
British journal of anaesthesia (2014-06-21)
P Lirk, M W Hollmann, M Fleischer, N C Weber, H Fiegl
ABSTRACT
Lidocaine demethylates deoxyribonucleic acid (DNA) in breast cancer cells. This modification of epigenetic information may be of therapeutic relevance in the perioperative period, because a decrease in methylation can reactivate tumour suppressor genes and inhibit tumour growth. The objectives of this study were to determine the effect of two amide local anaesthetics, ropivacaine and bupivacaine, on methylation in two breast cancer cell lines and to detect whether the combination of lidocaine with the chemotherapy agent 5-aza-2'-deoxycytidine (DAC) would result in additive demethylating effects. Breast cancer cell lines BT-20 [oestrogen receptor (ER)-negative] and MCF-7 (ER-positive) were incubated with lidocaine, bupivacaine, and ropivacaine to assess demethylating properties. Then, we tested varying concentrations of lidocaine and DAC to assess whether their demethylating effects were additive. Cell numbers and global methylation status were analysed. Lidocaine decreased methylation in BT-20 and MCF-7 cells, ropivacaine decreased methylation in BT-20 cells, and bupivacaine had no demethylating effect. When combined, lidocaine and DAC had additive demethylating effects. At clinically relevant doses, lidocaine and ropivacaine exert demethylating effects on specific breast cancer cell lines, but bupivacaine does not. The demethylating effects of lidocaine and DAC are indeed additive.
Sigma-Aldrich
Ropivacaine hydrochloride monohydrate, ≥98% (HPLC)
Pricing and availability is not currently available.