Merck
  • Cholesterol is Inefficiently Converted to Cholesteryl Esters in the Blood of Cardiovascular Disease Patients.
Cholesterol is Inefficiently Converted to Cholesteryl Esters in the Blood of Cardiovascular Disease Patients.
Scientific reports (2018-10-05)
Mathias J Gerl, Winchil L C Vaz, Neuza Domingues, Christian Klose, Michal A Surma, Júlio L Sampaio, Manuel S Almeida, Gustavo Rodrigues, Pedro Araújo-Gonçalves, Jorge Ferreira, Claudia Borbinha, João Pedro Marto, Miguel Viana-Baptista, Kai Simons, Otilia V Vieira
ABSTRACT
Shotgun lipidomic analysis of 203 lipids in 13 lipid classes performed on blood plasma of donors who had just suffered an acute coronary syndrome (ACS, n = 74), or an ischemic stroke (IS, n = 21), or who suffer from stable angina pectoris (SAP, n = 78), and an age-matched control cohort (n = 52), showed some of the highest inter-lipid class correlations between cholesteryl esters (CE) and phosphatidylcholines (PC) sharing a common fatty acid. The concentration of lysophospatidylcholine (LPC) and ratios of concentrations of CE to free cholesterol (Chol) were also lower in the CVD cohorts than in the control cohort, indicating a deficient conversion of Chol to CE in the blood plasma in the CVD subjects. A non-equilibrium reaction quotient, Q', describing the global homeostasis of cholesterol as manifested in the blood plasma was shown to have a value in the CVD cohorts (Q'ACS = 0.217 ± 0.084; Q'IS = 0.201 ± 0.084; Q'SAP = 0.220 ± 0.071) that was about one third less than in the control cohort (Q'Control = 0.320 ± 0.095, p < 1 × 10-4), suggesting its potential use as a rapid predictive/diagnostic measure of CVD-related irregularities in cholesterol homeostasis.
Product Description
Avanti
15:0 cholesteryl-d7 ester, Avanti Research - A Croda Brand 700144P, powder
Pricing and availability is not currently available.