Merck
  • Transcriptome Analysis Identifies ALCAM Overexpression as a Prognosis Biomarker in Laryngeal Squamous Cell Carcinoma.
Transcriptome Analysis Identifies ALCAM Overexpression as a Prognosis Biomarker in Laryngeal Squamous Cell Carcinoma.
Cancers (2020-02-23)
Pedro Nicolau-Neto, Paulo Thiago de Souza-Santos, Mariana Severo Ramundo, Priscila Valverde, Ivanir Martins, Izabella Costa Santos, Fernando Dias, Tatiana de Almeida Simão, Luis Felipe Ribeiro Pinto
ABSTRACT
Laryngeal squamous cell carcinoma (LSCC) is one of the most incident tumors in the world, especially in developing countries, such as Brazil. Different from other tumors, LSCC prognosis did not improve during the past four decades. Therefore, the objective of this study was to develop biomarkers that can predict LSCC patient's prognosis. Transcriptome analysis pointed out 287 overexpressed genes in LSCC in comparison to adjacent mucosa. Among these, a gene-pattern signature was created with 24 genes associated with prognosis. The Bayesian clustering of both Brazil and The Cancer Genome Atlas (TCGA) data pointed out clusters of samples possessing significative differences in the prognosis, and the expression panel of three genes (ALCAM, GBP6, and ME1) was capable to distinguish patients with worse prognosis with an accuracy of 97%. Survival analyses with TCGA data highlighted ALCAM gene expression as an independent prognostic factor for LSCC. This was further confirmed through immunohistochemistry, using a validation set of Brazilian patients. ALCAM expression was not associated with prognosis for other head and neck tumor sites. ALCAM overexpression seems to be an independent prognosis biomarker for LSCC patients.
Sigma-Aldrich
Anti-ALCAM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.