Merck
  • Phosphoglycerate Mutase 1 Activates DNA Damage Repair via Regulation of WIP1 Activity.
Phosphoglycerate Mutase 1 Activates DNA Damage Repair via Regulation of WIP1 Activity.
Cell reports (2020-04-16)
Shigeo Ohba, Tor-Christian Aase Johannessen, Kamalakar Chatla, Xiaodong Yang, Russell O Pieper, Joydeep Mukherjee
ABSTRACT
The metabolic enzyme phosphoglycerate mutase 1 (PGAM1) is overexpressed in several types of cancer, suggesting an additional function beyond its established role in the glycolytic pathway. We here report that PGAM1 is overexpressed in gliomas where it increases the efficiency of the DNA damage response (DDR) pathway by cytoplasmic binding of WIP1 phosphatase, thereby preventing WIP1 nuclear translocation and subsequent dephosphorylation of the ATM signaling pathway. Silencing of PGAM1 expression in glioma cells consequently decreases formation of γ-H2AX foci, increases apoptosis, and decreases clonogenicity following irradiation (IR) and temozolomide (TMZ) treatment. Furthermore, mice intracranially implanted with PGAM1-knockdown cells have significantly improved survival after treatment with IR and TMZ. These effects are counteracted by exogenous expression of two kinase-dead PGAM1 mutants, H186R and Y92F, indicating an important non-enzymatic function of PGAM1. Our findings identify PGAM1 as a potential therapeutic target in gliomas.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
PP1, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human PPM1D