Merck
  • A rapid and economical method of preparing radioiodinated cyclic nucleotide derivatives for use in radioimmunoassays.
A rapid and economical method of preparing radioiodinated cyclic nucleotide derivatives for use in radioimmunoassays.
Journal of immunoassay (1990-01-01)
W F Oehlenschlager, S W Kubalak, M G Currie
ABSTRACT
2'-O-succinyladenosine 3':5'-cyclic monophosphate tyrosyl methyl ester (ScAMP-TME) and 2'-O-succinylguanosine 3':5'-cyclic monophosphate tyrosyl methyl ester (ScGMP-TME) were radioiodinated using chloramine T and Na125I. The resulting radiolabeled cyclic nucleotide derivatives, ScAMP-125I-TME and ScGMP-125I-TME, were subsequently purified by reverse-phase chromatography on Sep-Pak C18 cartridges (Waters Associates, Milford, MA) and tested as tracers in sensitive radioimmunoassays for cAMP and cGMP, respectively. Purified ScAMP-125I-TME and ScGMP-125I-TME functioned in the respective radioimmunoassays for up to 12 weeks when suspended in a 1:1 (v:v) mixture of n-propanol and 20 mM sodium acetate, pH 6.0. Thus, this purification method enables rapid and economical preparation of tracers for cyclic nucleotide radioimmunoassays. Furthermore, our findings suggest that reverse-phase chromatography may be applicable to the purification of other small polar molecules to which tyrosyl groups have been added for the purpose of radioiodination.