Merck
  • Revisiting intrinsic transcription termination in mycobacteria: U-tract downstream of secondary structure is dispensable for termination.
Revisiting intrinsic transcription termination in mycobacteria: U-tract downstream of secondary structure is dispensable for termination.
Biochemical and biophysical research communications (2019-11-25)
Ezaz Ahmad, Shubhada R Hegde, Valakunja Nagaraja
ABSTRACT
In Escherichia coli, G/C-rich hairpin structure followed by a U-tract in the 3' region of the nascent RNA are crucial determinants for intrinsic or factor independent transcription termination. In mycobacteria, there is a scarcity of such intrinsic terminators. However, secondary structures having G/C-rich stem devoid of any U's or with suboptimal U-tracts were identified earlier as terminators and found to be functional both in vitro and in vivo. Two different observations - that a mycobacterial RNA polymerase (RNAP) does not function at intrinsic terminators devoid of U-tracts and the identification of an altogether new motif for termination in mycobacteria necessitated re-examining a number of putative terminators for their function as terminators. When these in silico identified non-canonical terminators were subjected to experimental validation, they were found to dissociate RNA from the elongating RNAP. Termination is observed when the U-tracts were reduced, or totally absent both in vitro and in vivo. Our results, thus indicate that the presence of U-tract following the G/C-rich stem in an intrinsic terminator may not be an essential determinant for transcription termination in mycobacteria.
Sigma-Aldrich
(±)-Geosmin, ≥97% (GC)
Pricing and availability is not currently available.