Merck
  • Systems biology analysis of Drosophila in vivo screen data elucidates core networks for DNA damage repair in SCA1.
Systems biology analysis of Drosophila in vivo screen data elucidates core networks for DNA damage repair in SCA1.
Human molecular genetics (2013-11-02)
Sam S Barclay, Takuya Tamura, Hikaru Ito, Kyota Fujita, Kazuhiko Tagawa, Teppei Shimamura, Asuka Katsuta, Hiroki Shiwaku, Masaki Sone, Seiya Imoto, Satoru Miyano, Hitoshi Okazawa
ABSTRACT
DNA damage repair is implicated in neurodegenerative diseases; however, the relative contributions of various DNA repair systems to the pathology of these diseases have not been investigated systematically. In this study, we performed a systematic in vivo screen of all available Drosophila melanogaster homolog DNA repair genes, and we tested the effect of their overexpression on lifespan and developmental viability in Spinocerebellar Ataxia Type 1 (SCA1) Drosophila models expressing human mutant Ataxin-1 (Atxn1). We identified genes previously unknown to be involved in CAG-/polyQ-related pathogenesis that function in multiple DNA damage repair systems. Beyond the significance of each repair system, systems biology analyses unraveled the core networks connecting positive genes in the gene screen that could contribute to SCA1 pathology. In particular, RpA1, which had the largest effect on lifespan in the SCA1 fly model, was located at the hub position linked to such core repair systems, including homologous recombination (HR). We revealed that Atxn1 actually interacted with RpA1 and its essential partners BRCA1/2. Furthermore, mutant but not normal Atxn1 impaired the dynamics of RpA1 in the nucleus after DNA damage. Uptake of BrdU by Purkinje cells was observed in mutant Atxn1 knockin mice, suggesting their abnormal entry to the S-phase. In addition, chemical and genetic inhibitions of Chk1 elongated lifespan and recovered eye degeneration. Collectively, we elucidated core networks for DNA damage repair in SCA1 that might include the aberrant usage of HR.
Sigma-Aldrich
Anti-Ataxin-1 Antibody, 11NQ, clone N76/8, clone N76/8, from mouse
Sigma-Aldrich
Anti-Polyglutamine-Expansion Diseases Marker Antibody, clone 5TF1-1C2, ascites fluid, clone 5TF1-1C2, Chemicon®