Merck
  • SLMP53-1 Inhibits Tumor Cell Growth through Regulation of Glucose Metabolism and Angiogenesis in a P53-Dependent Manner.
SLMP53-1 Inhibits Tumor Cell Growth through Regulation of Glucose Metabolism and Angiogenesis in a P53-Dependent Manner.
International journal of molecular sciences (2020-01-23)
Helena Ramos, Juliana Calheiros, Joana Almeida, Valentina Barcherini, Sónia Santos, Alexandra T P Carvalho, Maria M M Santos, Lucília Saraiva
ABSTRACT
The Warburg effect is an emerging hallmark of cancer, which has the tumor suppressor p53 as its major regulator. Herein, we unveiled that p53 activation by (S)-tryptophanol-derived oxazoloisoindolinone (SLMP53-1) mediated the reprograming of glucose metabolism in cancer cells and xenograft human tumor tissue, interfering with angiogenesis and migration. Particularly, we showed that SLMP53-1 regulated glycolysis by downregulating glucose transporter 1 (GLUT1), hexokinase-2 (HK2), and phosphofructokinase-2 isoform 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-3 (PFKFB3) (key glycolytic enzymes), while upregulating the mitochondrial markers synthesis of cytochrome c oxidase 2 (SCO2), cytochrome c oxidase subunit 4 (COX4), and OXPHOS mitochondrial complexes. SLMP53-1 also downregulated the monocarboxylate transporter 4 (MCT4), causing the subsequent reduction of lactate export by cancer cells. Besides the acidification of the extracellular environment, SLMP53-1 further increased E-cadherin and reduced metalloproteinase-9 (MMP-9) expression levels in both cancer cells and xenograft human tumor tissue, which suggested the interference of SLMP53-1 in extracellular matrix remodeling and epithelial-to-mesenchymal transition. Consistently, SLMP53-1 depleted angiogenesis, decreasing endothelial cell tube formation and vascular endothelial growth factor (VEGF) expression levels. SLMP53-1 also exhibited synergistic growth inhibitory activity in combination with the metabolic modulator dichloroacetic acid. These data reinforce the promising application of the p53-activating agent SLMP53-1 in cancer therapy, by targeting p53-mediated pathways of growth and dissemination.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-HK2 Antibody, clone 3D3, ascites fluid, clone 3D3, from mouse
Pricing and availability is not currently available.