Merck
  • Structure-Function Studies on IMD-0354 Identifies Highly Active Colistin Adjuvants.
Structure-Function Studies on IMD-0354 Identifies Highly Active Colistin Adjuvants.
ChemMedChem (2019-11-23)
Ansley M Nemeth, Akash K Basak, Alexander W Weig, Santiana A Marrujo, William T Barker, Leigh A Jania, Tyler A Hendricks, Ashley E Sullivan, Patrick M O'Connor, Roberta J Melander, Beverly H Koller, Christian Melander
ABSTRACT
Infections caused by multidrug-resistant (MDR) bacteria, particularly Gram-negative bacteria, are an escalating global health threat. Often clinicians are forced to administer the last-resort antibiotic colistin; however, colistin resistance is becoming increasingly prevalent, giving rise to the potential for a situation in which there are no treatment options for MDR Gram-negative infections. The development of adjuvants that circumvent bacterial resistance mechanisms is a promising orthogonal approach to the development of new antibiotics. We recently disclosed that the known IKK-β inhibitor IMD-0354 potently suppresses colistin resistance in several Gram-negative strains. In this study, we explore the structure-activity relationship (SAR) between the IMD-0354 scaffold and colistin resistance suppression, and identify several compounds with more potent activity than the parent against highly colistin-resistant strains of Acinetobacter baumannii and Klebsiella pneumoniae.
Sigma-Aldrich
Colistin sulfate salt, ≥19,000 IU/mg
Pricing and availability is not currently available.