Merck
  • Conversion of D-fructose to 5-acetoxymethyl-2-furfural Using Immobilized Lipase and Cation Exchange Resin.
Conversion of D-fructose to 5-acetoxymethyl-2-furfural Using Immobilized Lipase and Cation Exchange Resin.
Molecules (Basel, Switzerland) (2019-12-22)
Nhan Thanh Thien Huynh, Kyung Won Lee, Jin Ku Cho, Yong Jin Kim, Se Won Bae, Jong Shik Shin, Seunghan Shin
ABSTRACT
5-Acetoxymethyl-2-furfural (AMF) was prepared from D-fructose via 1,6-diacetylfructose (DAF) through a simple two-step reaction pathway. Immobilized enzyme (Novozym 435) was found to be the best enzymatic catalyst for the trans-esterification step (yielding 94.6% DAF). In the dehydration step, while soluble H2SO4 was found to be the best acidic catalyst (yielding 86.6% AMF), we opted to utilize heterogeneous cation exchange resin (Amberlyst 15) together with recyclable industrial solvents (1,4-dioxane) for a more sustainable AMF synthesis procedure. Although the total yield of AMF was a little lower, both the enzyme and the solid acid catalyst could be recycled for five cycles without a significant loss of activity, which has a major contribution to the cost-efficient aspect of the entire process.
Sigma-Aldrich
Vinyl acetate, contains 3-20 ppm hydroquinone as inhibitor, ≥99%