Merck
  • Comparative study on enantiomer resolving ability of amylose tris(3-chloro-5-methylphenylcarbamate) covalently immobilized onto silica in nano-liquid chromatography and capillary electrochromatography.
Comparative study on enantiomer resolving ability of amylose tris(3-chloro-5-methylphenylcarbamate) covalently immobilized onto silica in nano-liquid chromatography and capillary electrochromatography.
Journal of chromatography. A (2019-09-01)
Giovanni D'Orazio, Chiara Fanali, Salvatore Fanali, Alessandra Gentili, Bezhan Chankvetadze
ABSTRACT
In the present study separation of enantiomers of some chiral neutral and weakly acidic analytes was investigated on the chiral stationary phase (CSP) made by covalent immobilization of amylose tris(3-chloro-5-methylphenylcarbamate) onto silica in nano-liquid chromatography (nano-LC) and capillary electrochromatography (CEC) in acetonitrile and aqueous acetonitrile. Few comparisons were made also between the enantioseparations in nano-LC and high-performance liquid chromatography (HPLC) with the chiral column of 4.6 × 250 mm dimension. Slightly better separation of enantiomers was observed in HPLC mode compared to nano-LC mode. It was shown that in the capillary columns packed with the CSP containing about 20% (w/w) of a covalently immobilized neutral chiral selector, amylose tris(3-chloro-5-methylphenylcarbamate), sufficient electroosmotic flow has been generated and enantioseparations with reasonable analysis time were performed also in CEC mode. It was shown once again that CEC offers a clear advantage over nano-LC from the viewpoint of plate numbers and peak resolution.
Product Description
Supelco
Dichlorprop, PESTANAL®, analytical standard
Sigma-Aldrich
6-Hydroxyflavanone, 99%
Supelco
Haloxyfop, PESTANAL®, analytical standard
Sigma-Aldrich
7-Hydroxyflavanone, 98%