Merck
  • Vascular Adhesion Protein-1 (VAP-1) as Predictor of Radiographic Severity in Symptomatic Knee Osteoarthritis in the New York University Cohort.
Vascular Adhesion Protein-1 (VAP-1) as Predictor of Radiographic Severity in Symptomatic Knee Osteoarthritis in the New York University Cohort.
International journal of molecular sciences (2019-05-31)
Eirini Bournazou, Jonathan Samuels, Hua Zhou, Svetlana Krasnokutsky, Jyoti Patel, Tianzhen Han, Jenny Bencardino, Leon Rybak, Steven B Abramson, Uwe Junker, Karen S Brown, Mukundan Attur
ABSTRACT
To investigate the expression of vascular adhesion protein-1 (VAP-1) in joint tissues and serum in symptomatic knee osteoarthritis (SKOA) patients and examine whether VAP-1 levels predict increased risk of disease severity in a cross-sectional study. Baseline VAP-1 expression and soluble VAP-1 (sVAP-1) levels were assessed in the synovium synovial fluid and in the serum in cohorts of patients with tibiofemoral medial knee OA and healthy subjects. Standardized fixed-flexion poster anterior knee radiographs scored for Kellgren-Lawrence (KL) grade (0-4) and medial joint space width (JSW). KL1/2 vs. KL3/4 scores defined early and advanced radiographic severity, respectively. Biochemical markers assessed in serum or synovial fluids (SF) comprised sVAP-1, interleukin 1 receptor antagonist (IL-1Ra), interleukin 6 (IL-6), soluble receptor for advanced glycation end-products (sRAGE), C-C motif chemokine ligand 2 (CCL2), C-C motif chemokine ligand 4 (CCL4), cluster of differentiation 163 (CD163), high sensitivity C-reactive protein (hsCRP), and matrix metalloproteinases (MMPs)-1,-3,-9. Associations between biomarkers and radiographic severity KL1/2 vs. KL3/4 (logistic regression controlling for covariates) and pain (Spearman correlation) were evaluated. Elevated levels of sVAP-1 observed in OA synovial fluid and VAP-1 expression in synovium based on immunohistochemical, microarray, and real-time quantitative polymerase chain reaction (qRT-PCR) analyses. However, serum sVAP-1 levels in OA patients were lower than in controls and inversely correlated with pain and inflammation markers (hsCRP and soluble RAGE). Soluble VAP-1 levels in serum were also lower in radiographically advanced (KL3/4) compared with early KL1/2 knee SKOA patients. Local (synovial fluid) semicarbazide-sensitive amine oxidase (SSAO)/sVAP-1 levels were elevated in OA and correlated with radiographic severity. However, systemic (serum) sVAP-1 levels were lower in SKOA patients than normal and inversely correlated with pain and inflammation markers. Serum sVAP-1 levels were higher in early (KL1/2) compared with advanced (KL3/4) SKOA patients.
Sigma-Aldrich
Anti-AOC3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution