Merck
  • The Precursor for Nerve Growth Factor (proNGF) in Thyroid Cancer Lymph Node Metastases: Correlation with Primary Tumour and Pathological Variables.
The Precursor for Nerve Growth Factor (proNGF) in Thyroid Cancer Lymph Node Metastases: Correlation with Primary Tumour and Pathological Variables.
International journal of molecular sciences (2019-11-30)
Christopher W Rowe, Tony Dill, Sam Faulkner, Craig Gedye, Jonathan W Paul, Jorge M Tolosa, Mark Jones, Simon King, Roger Smith, Hubert Hondermarck
ABSTRACT
Metastases in thyroid cancer are associated with aggressive disease and increased patient morbidity, but the factors driving metastatic progression are unclear. The precursor for nerve growth factor (proNGF) is increased in primary thyroid cancers, but its expression or significance in metastases is not known. In this study, we analysed the expression of proNGF in a retrospective cohort of thyroid cancer lymph node metastases (n = 56), linked with corresponding primary tumours, by automated immunohistochemistry and digital quantification. Potential associations of proNGF immunostaining with clinical and pathological parameters were investigated. ProNGF staining intensity (defined by the median h-score) was significantly higher in lymph node metastases (h-score 94, interquartile range (IQR) 50-147) than in corresponding primary tumours (57, IQR 42-84) (p = 0.002). There was a correlation between proNGF expression in primary tumours and corresponding metastases, where there was a 0.68 (95% CI 0 to 1.2) increase in metastatic tumour h-score for each unit increase in the primary tumour h-score. However, larger tumours (both primary and metastatic) had lower proNGF expression. In a multivariate model, proNGF expression in nodal metastases was negatively correlated with lateral neck disease and being male. In conclusion, ProNGF is expressed in locoregional metastases of thyroid cancer and is higher in lymph node metastases than in primary tumours, but is not associated with high-risk clinical features.
Sigma-Aldrich
Anti-Nerve Growth Factor Antibody, pro, serum, Chemicon®
Pricing and availability is not currently available.