Merck
  • A novel sphingomyelin/cholesterol domain-specific probe reveals the dynamics of the membrane domains during virus release and in Niemann-Pick type C.
A novel sphingomyelin/cholesterol domain-specific probe reveals the dynamics of the membrane domains during virus release and in Niemann-Pick type C.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2016-08-06)
Asami Makino, Mitsuhiro Abe, Reiko Ishitsuka, Motohide Murate, Takuma Kishimoto, Shota Sakai, Françoise Hullin-Matsuda, Yukiko Shimada, Takehiko Inaba, Hideyuki Miyatake, Hideko Tanaka, Atsushi Kurahashi, Chan-Gi Pack, Rinshi S Kasai, Shuku Kubo, Nicole L Schieber, Naoshi Dohmae, Naoya Tochio, Kyoji Hagiwara, Yutaka Sasaki, Yoko Aida, Fumihiro Fujimori, Takanori Kigawa, Kozo Nishibori, Robert G Parton, Akihiro Kusumi, Yasushi Sako, Gregor Anderluh, Makoto Yamashita, Tetsuyuki Kobayashi, Peter Greimel, Toshihide Kobayashi
ABSTRACT
We identified a novel, nontoxic mushroom protein that specifically binds to a complex of sphingomyelin (SM), a major sphingolipid in mammalian cells, and cholesterol (Chol). The purified protein, termed nakanori, labeled cell surface domains in an SM- and Chol-dependent manner and decorated specific lipid domains that colocalized with inner leaflet small GTPase H-Ras, but not K-Ras. The use of nakanori as a lipid-domain-specific probe revealed altered distribution and dynamics of SM/Chol on the cell surface of Niemann-Pick type C fibroblasts, possibly explaining some of the disease phenotype. In addition, that nakanori treatment of epithelial cells after influenza virus infection potently inhibited virus release demonstrates the therapeutic value of targeting specific lipid domains for anti-viral treatment.-Makino, A., Abe, M., Ishitsuka, R., Murate, M., Kishimoto, T., Sakai, S., Hullin-Matsuda, F., Shimada, Y., Inaba, T., Miyatake, H., Tanaka, H., Kurahashi, A., Pack, C.-G., Kasai, R. S., Kubo, S., Schieber, N. L., Dohmae, N., Tochio, N., Hagiwara, K., Sasaki, Y., Aida, Y., Fujimori, F., Kigawa, T., Nishibori, K., Parton, R. G., Kusumi, A., Sako, Y., Anderluh, G., Yamashita, M., Kobayashi, T., Greimel, P., Kobayashi, T. A novel sphingomyelin/cholesterol domain-specific probe reveals the dynamics of the membrane domains during virus release and in Niemann-Pick type C.
Sigma-Aldrich
Anti-Influenza A Antibody, H1N1 Antigen, clone 9B3.2, ascites fluid, clone 9B3.2, Chemicon®
Pricing and availability is not currently available.