Merck
  • Neural stem cells for disease modeling and evaluation of therapeutics for infantile (CLN1/PPT1) and late infantile (CLN2/TPP1) neuronal ceroid lipofuscinoses.
Neural stem cells for disease modeling and evaluation of therapeutics for infantile (CLN1/PPT1) and late infantile (CLN2/TPP1) neuronal ceroid lipofuscinoses.
Orphanet journal of rare diseases (2018-04-11)
Ni Sima, Rong Li, Wei Huang, Miao Xu, Jeanette Beers, Jizhong Zou, Steven Titus, Elizabeth A Ottinger, Juan J Marugan, Xing Xie, Wei Zheng
ABSTRACT
Infantile and late infantile neuronal ceroid lipofuscinoses (NCLs) are lysosomal storage diseases affecting the central nervous system (CNS). The infantile NCL (INCL) is caused by mutations in the PPT1 gene and late-infantile NCL (LINCL) is due to mutations in the TPP1 gene. Deficiency in PPT1 or TPP1 enzyme function results in lysosomal accumulation of pathological lipofuscin-like material in the patient cells. There is currently no small-molecular drug treatment for NCLs. We have generated induced pluripotent stem cells (iPSC) from three patient dermal fibroblast lines and further differentiated them into neural stem cells (NSCs). Using these new disease models, we evaluated the effect of δ-tocopherol (DT) and hydroxypropyl-β-cyclodextrin (HPBCD) with the enzyme replacement therapy as the control. Treatment with the relevant recombinant enzyme or DT significantly ameliorated the lipid accumulation and lysosomal enlargement in the disease cells. A combination therapy of δ-tocopherol and HPBCD further improved the effect compared to that of either drug used as a single therapy. The results demonstrate that these patient iPSC derived NCL NSCs are valid cell- based disease models with characteristic disease phenotypes that can be used for study of disease pathophysiology and drug development.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate, clone TRA-1-60, from mouse, FITC conjugate
Sigma-Aldrich
Mouse IgG2b Negative Control, clone GC198, FITC conjugate, Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.