Merck
  • New family with HSPB8-associated autosomal dominant rimmed vacuolar myopathy.
New family with HSPB8-associated autosomal dominant rimmed vacuolar myopathy.
Neurology. Genetics (2019-08-14)
Sejad Al-Tahan, Lan Weiss, Howard Yu, Sha Tang, Mario Saporta, Anna Vihola, Tahseen Mozaffar, Bjarne Udd, Virginia Kimonis
ABSTRACT
We clinically and molecularly characterize a new family with autosomal dominant rimmed vacuolar myopathy (RVM) caused by mutations in the HSPB8 gene. We performed whole-exome and whole-genome sequencing in the family. Western blot and immunocytochemistry were used to analyze 3 patient fibroblasts, and findings were compared with their age- and sex-matched controls. Affected patients have distal and proximal myopathy, with muscle biopsy showing rimmed vacuoles, muscle fiber atrophy, and endomysial fibrosis typical of RVM. Muscle MRI showed severe relatively symmetric multifocal fatty degenerative changes of the lower extremities. We identified a duplication of C at position 515 of the HSPB8 gene (c.515dupC) by whole-genome sequencing, which caused a frameshift with a predicted alternate stop codon p.P173SFS*43 in all affected individuals, resulting in an elongated protein product. Western blot and immunocytochemistry studies revealed reduced expression of heat shock protein beta 8 in patient fibroblasts compared with control fibroblasts, in addition to disrupted autophagy pathology. We report a novel family with autosomal dominant RVM caused by the c.515dupC mutation of the HSPB8 gene, causing a translational frameshift that results in an elongated protein. Understanding the mechanism for the RVM pathology caused by mutated chaperone will permit novel targeted strategies to alter the natural history progression. As next-generation sequencing becomes more available, additional myopathic families will be identified with HSPB8 mutations.
Sigma-Aldrich
Anti-HSPB8 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.