- Effects of IL-10- and FasL-overexpressing dendritic cells on liver transplantation tolerance in a heterotopic liver transplantation rat model.
Effects of IL-10- and FasL-overexpressing dendritic cells on liver transplantation tolerance in a heterotopic liver transplantation rat model.
Acute rejection is the major determinant for the long-term survival of donor liver after liver transplantation (LT). The aim of this study was to examine the therapeutic potential of interleukin (IL)-10-FasL-overexpressing immature dendritic cells (imDCs) to induce local immunosuppression in liver grafts. imDCs derived from donors were transduced by lentiviral vectors expressing human IL-10 and/or Fas ligand (FasL) gene(s), and the expression of surface molecules and the ability to induce T-cell proliferation were measured. imDCs were intraperitoneally injected into recipient rats as a model of LT to examine the rejection grade [Banff rejection activity index (RAI)], liver functions [Alanine aminotransferase, Aspartate aminotransferase (AST) and total bilirubin (TBIL)] and post-transplant survival. IL-10 and FasL co-transduction of imDCs induced a greater reduction in CD80, CD86 and major histocompatibility complex class II (MHC II) expression, as well as T-cell proliferation, but increased levels of IL-10 and FasL in culture supernatants compared with mono-transduced or untransduced imDCs (P < 0.05). The infusion of co-transduced imDCs in LT recipients reduced RAI scores, decreased plasma AST and TBIL, and prolonged survival compared with mono-transduced or untransduced imDC-treated liver allografts. These findings demonstrated that the transfusion of IL-10-FasL/imDCs enhanced immune tolerance and prolonged the survival of liver allografts after LT. The immunomodulatory activity of IL-10- and FasL-modified imDCs might be a new therapeutic approach to prevent organ rejection in clinical transplantation.